Learn More
4-(Hydroxymethyl)pyridine-2-carboxylic acid, 95%
Product Code. 15495697
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 15495697
Brand: Thermo Scientific Alfa Aesar H32921.14
Request Bulk or Custom Format
CAS: 923169-37-3 | C7H7NO3 | 153.137 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 923169-37-3
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD08447083
InChI Key QMLYYPGJXCVEHA-UHFFFAOYSA-N
Synonym 4-hydroxymethyl picolinic acid, 4-hydroxymethyl pyridine-2-carboxylic acid, 4-hydroxymethyl pyridine-2-carboxylicacid
PubChem CID 22645434
IUPAC Name 4-(hydroxymethyl)pyridine-2-carboxylic acid
SMILES C1=CN=C(C=C1CO)C(=O)O
Specifications
Melting Point 178°C to 185°C (decomposition)
Quantity 25 g
Formula Weight 153.15
Percent Purity 95%
Chemical Name or Material 4-(Hydroxymethyl)pyridine-2-carboxylic acid