Learn More
4-Hydroxybenzenesulfonic Acid (85%), TRC
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzenesulfonic Acid |
| CAS | 98-67-9 |
| Molecular Formula | C6H6O4S |
| Quantity | 50 g |
| Synonym | 4-Hydroxyphenylsulfonic acid, 4-Phenolsulfonic acid, NSC 227908, p-Hydroxybenzenesulfonic acid, p-Phenolsulfonic acid, p-Sulfophenol |
| InChI Formula | InChI=1S/C6H6O4S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4,7H,(H,8,9,10) |
| SMILES | CCC(C(N)=O)(C(N)=O)c1ccccc1 |
| IUPAC Name | 4-hydroxybenzenesulfonic acid |
| Molecular Weight (g/mol) | 174.17 |
| Formula Weight | 173.9987 |