Learn More
4-Hydroxyantipyrine, TRC
Product Code. 17990885
Change view
Click to view available options
Quantity:
25 mg
250 mg
500 mg
Unit Size:
250mg
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17990885 250 mg 250mg
17900735 500 mg 500mg
17983125 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17990885 Supplier Toronto Research Chemicals Supplier No. TRCH801805250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1672-63-5
Molecular Formula C11 H12 N2 O2
Molecular Weight (g/mol) 204.2252
Synonym Antipyrine, 4-hydroxy- (6CI,7CI,8CI), 1,2-Dihydro-4-hydroxy-1,5-dimethyl-2-phenyl-3H-pyrazol-3-one, 4-Hydroxy-1,5-dimethyl-3-oxo-2-phenyl pyrazoline, 4-Hydroxyantipyrine, 4-Hydroxyphenazone, NSC 174055
IUPAC Name 4-hydroxy-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CN1N(C(=O)C(=C1C)O)c2ccccc2
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C11H12N2O2/c1-8-10(14)11(15)13(12(8)2)9-6-4-3-5-7-9/h3-7,14H,1-2H3
Formula Weight 204.0899
Chemical Name or Material 4-Hydroxyantipyrine