Learn More
4-Hydroxy-3,5-dimethoxybenzhydrazide, 97%, Thermo Scientific™
Product Code. 11313156
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313156 5 g 5g
11303156 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11313156 Supplier Thermo Scientific Alfa Aesar Supplier No. L11032.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1443-76-1
Molecular Formula C9H12N2O4
Molecular Weight (g/mol) 212.205
MDL Number MFCD00017077
InChI Key AKZAAANGIPRVDU-UHFFFAOYSA-N
Synonym 3,5-dimethoxy-4-hydroxybenzhydrazide, 4-hydroxy-3,5-dimethoxybenzhydrazide, benzoic acid, 4-hydroxy-3,5-dimethoxy-, hydrazide, acmc-1bzfx, 4-???-2,6-dimethoxyphenol, #, 3,5-dimethoxy-4-hydroxy benzhydrazide, 4-hydroxy-3,5-dimethoxy-benzoic acid hydrazide, benzoic acid,4-hydroxy-3,5-dimethoxy-, hydrazide
PubChem CID 74043
IUPAC Name 4-hydroxy-3,5-dimethoxybenzohydrazide
SMILES COC1=CC(=CC(=C1O)OC)C(=O)NN
Specifications
Melting Point 154°C to 157°C
Quantity 5 g
Beilstein 2657142
Formula Weight 212.21
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3,5-dimethoxybenzhydrazide