missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-2-hexenal Antibody (5C11.1) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18263JF585
This item is not returnable.
View return policy
Description
4-Hydroxy-2-hexenal Monoclonal antibody specifically detects 4-Hydroxy-2-hexenal in Nonspecies specific samples. It is validated for ELISA,Western Blot,Dot Blot
Specifications
| 4-Hydroxy-2-hexenal | |
| Monoclonal | |
| Janelia Fluor 585 | |
| 4-HHE;4-hydroxy Hexenal;4-Hydroxy-2-hexenal;4-Hydroxy-2-hexenal-modified;HHE;MG;MG-modified protein | |
| Synthetic 4-Hydroxy-2-hexenal modified Keyhole Limpet Hemocyanin (KLH) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Western Blot, Dot Blot | |
| 5C11.1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction