missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzylamine, 98+%, Thermo Scientific Chemicals

Catalog Number p-7022620
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
140-75-0
C7H8FN
125.146
MFCD00008120
IIFVWLUQBAIPMJ-UHFFFAOYSA-N
4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392
67326
(4-fluorophenyl)methanamine
C1=CC(=CC=C1CN)F
This item is not returnable. View return policy
140-75-0
C7H8FN
125.146
MFCD00008120
IIFVWLUQBAIPMJ-UHFFFAOYSA-N
4-fluorobenzylamine, 4-fluorophenyl methanamine, benzenemethanamine, 4-fluoro, p-fluorobenzylamine, benzylamine, p-fluoro, p-fluorobenzyl amine, 4-fluoro-benzylamine, 1-4-fluorophenyl methanamine, 4-fluorobenzyl amine, chembl12392
67326
(4-fluorophenyl)methanamine
C1=CC(=CC=C1CN)F

4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Fluorobenzylamine is used as a potassium channel blocking agent. It is used in the synthesis of new tris-iron(III) chelates of 3-hydroxy-4-pyridinone ligands. It reacts with the α- and γ-carboxyl groups of folic acid to yield 18F-labeled folate2. It is an important building block for the synthesis of 18F-labeled compounds.

Solubility
Soluble in water.

Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agent, air, carbon dioxide.
TRUSTED_SUSTAINABILITY
Density 1.095
Boiling Point 182°C to 183°C
Flash Point 66°C (150°F)
Odor Amine-like
Refractive Index 1.513
Quantity 10 g
UN Number UN2735
Beilstein 636501
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 125.15
Percent Purity ≥98%
Chemical Name or Material 4-Fluorobenzylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'