Learn More
4-Fluoro-DL-phenylglycine, 97%, Thermo Scientific™
Product Code. 11319555
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11319555 1 g 1g
11329555 5 g 5g
2 options
Product Code. 11319555
Brand: Thermo Scientific Alfa Aesar L18932.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7292-73-1
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.16
MDL Number MFCD00066444,MFCD00042727
InChI Key JKFYKCYQEWQPTM-UHFFFAOYNA-N
Synonym 2-amino-2-4-fluorophenyl acetic acid, 4-fluoro-dl-phenylglycine, 4-fluoro-dl-alpha-phenylglycine, dl-4-fluorophenylglycine, amino 4-fluorophenyl acetic acid, 4-fluorophenylglycine, dl-4-fluorophenyl glycine, amino-4-fluoro-phenyl-acetic acid, dl-alpha-4-fluorophenyl-glycine, benzeneacetic acid, alpha-amino-4-fluoro
PubChem CID 98313
IUPAC Name 2-amino-2-(4-fluorophenyl)acetic acid
SMILES NC(C(O)=O)C1=CC=C(F)C=C1
Specifications
Melting Point >300°C
Beilstein 2804849
Quantity 1 g
Formula Weight 169.16
Percent Purity 97%
Chemical Name or Material 4-Fluoro-DL-phenylglycine