missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-3-(trifluoromethoxy)aniline, 97%, Thermo Scientific™

Product Code. 15445577
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15445577 1 g 1g
15455577 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445577 Supplier Thermo Scientific Alfa Aesar Supplier No. H32810.03
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Fluoro-3-(trifluoromethyl)aniline is used in the synthesis of trifluoromethylated poly(phenylene oxide). It is used in the preparation of 2-[3-(4-fluoro-3-trifluoromethyl-phenyl)-ureido]-2-methyl-propionic acid methyl ester by reacting with 2-isocyanato-2-methyl-propionic acid methyl ester. It is also used as a chemical intermediate in the synthesis of agrochemicals, pharmaceuticals and dyes.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113421-98-0
Molecular Formula C7H5F4NO
Molecular Weight (g/mol) 195.12
MDL Number MFCD09832246
InChI Key JSMGZPOPIVVIRZ-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethoxy aniline, benzenamine, 4-fluoro-3-trifluoromethoxy, 4-fluoro-3-trifluoromethoxy-aniline, 4-fluoro-3-trifluoromethoxy phenylamine, 4-fluoro-3-trifluoro methoxy aniline
PubChem CID 21614378
IUPAC Name 4-fluoro-3-(trifluoromethoxy)aniline
SMILES NC1=CC(OC(F)(F)F)=C(F)C=C1

Specifications

Refractive Index 1.4502
Quantity 1 g
UN Number UN2810
Formula Weight 195.12
Percent Purity 97%
Chemical Name or Material 4-Fluoro-3-(trifluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.