Learn More
4-Fluoro-2-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
Product Code. 11474447
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11474447 1 g 1g
11484447 5 g 5g
2 options
Product Code. 11474447
Brand: Thermo Scientific Alfa Aesar B20469.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 202522-22-3
Molecular Formula C8H8F4N
Molecular Weight (g/mol) 194.15
MDL Number MFCD00061164
InChI Key BLQGRYYLPWCHMA-UHFFFAOYSA-O
PubChem CID 2734878
SMILES [NH3+]CC1=CC=C(F)C=C1C(F)(F)F
Specifications
Quantity 1 g
UN Number UN2735
Synonym 4-fluoro-2-trifluoromethyl benzylamine, 4-fluoro-2-trifluoromethyl phenyl methanamine, 1-4-fluoro-2-trifluoromethyl phenyl methanamine, 4-fluoro-2-trifluoromethyl benzyl amine, 4-fluoro-2-trifluoromethyl benzenemethanamine, 4-fluoro-2-trifluoromethylbenzylamine, benzenemethanamine,4-fluoro-2-trifluoromethyl, benzenemethanamine, 4-fluoro-2-trifluoromethyl, 4-fluoro-2-trifluoromethyl phenyl methylamine
IUPAC Name [4-fluoro-2-(trifluoromethyl)phenyl]methanamine
Formula Weight 193.15
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzylamine