missing translation for 'onlineSavingsMsg'
Learn More

4-Fluoro-2-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. p-7027629
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11460358 1 g 1g
11470358 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11460358

Brand: Thermo Scientific Alfa Aesar B24528.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206860-48-2
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.026
MDL Number MFCD00061176
InChI Key JMNOONULDANZRZ-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzyl bromide, 1-bromomethyl-4-fluoro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-fluoro-2-trifluoromethyl, alpha-bromo-4-fluoro-2-trifluoromethyl toluene, 1-bromomethyl-4-fluoro-2-trifluoromethyl-benzene, pubchem4642, acmc-20ai28, 4-fluoro-2-trifluoromethylbenzyl bromide, 4-fluoro-2-trifluoromethyl-benzyl bromide
PubChem CID 2737576
IUPAC Name 1-(bromomethyl)-4-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1F)C(F)(F)F)CBr

Specifications

Density 1.665
Boiling Point 141°C
Flash Point 85°C (185°F)
Refractive Index 1.4835
Quantity 1 g
UN Number UN1760
Sensitivity Moisture sensitive
Formula Weight 257.03
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.