Learn More
4-Fluoro-2-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™
Product Code. 11470358
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470358 5 g 5g
11460358 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11470358 Supplier Thermo Scientific Alfa Aesar Supplier No. B24528.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 206860-48-2
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.026
MDL Number MFCD00061176
InChI Key JMNOONULDANZRZ-UHFFFAOYSA-N
Synonym 4-fluoro-2-trifluoromethyl benzyl bromide, 1-bromomethyl-4-fluoro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl benzylbromide, benzene, 1-bromomethyl-4-fluoro-2-trifluoromethyl, alpha-bromo-4-fluoro-2-trifluoromethyl toluene, 1-bromomethyl-4-fluoro-2-trifluoromethyl-benzene, pubchem4642, acmc-20ai28, 4-fluoro-2-trifluoromethylbenzyl bromide, 4-fluoro-2-trifluoromethyl-benzyl bromide
PubChem CID 2737576
IUPAC Name 1-(bromomethyl)-4-fluoro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1F)C(F)(F)F)CBr
Specifications
Density 1.665
Boiling Point 141°C
Flash Point 85°C (185°F)
Refractive Index 1.4835
Quantity 5 g
UN Number UN1760
Sensitivity Moisture sensitive
Formula Weight 257.03
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzyl bromide