Learn More
4-Fluoro-2-iodobenzoic acid, 97%
Product Code. 15441828
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15441828 1 g 1g
15461828 250 mg 250mg
15451828 5 g 5g
3 options
Product Code. 15441828
Brand: Thermo Scientific Alfa Aesar H55531.03
Request Bulk or Custom Format
CAS: 56096-89-0 | C7H4FIO2 | 266.01 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56096-89-0
Molecular Formula C7H4FIO2
Molecular Weight (g/mol) 266.01
MDL Number MFCD04108278
InChI Key DUKFTVLJAXWGPI-UHFFFAOYSA-N
Synonym 4-fluoro-2-iodo-benzoic acid, 2-iodo-4-fluorobenzoic acid, 4-fluoro-2-iodobenzoicacid, benzoic acid, 4-fluoro-2-iodo, benzoic acid, 4-fluoroiodo, pubchem1389, acmc-1atr7, ksc494a9j, benzoic acid,4-fluoro-2-iodo, buttpark 83\07-33
PubChem CID 12520164
IUPAC Name 4-fluoro-2-iodobenzoic acid
SMILES C1=CC(=C(C=C1F)I)C(=O)O
Specifications
Melting Point 143°C to 147°C
Quantity 1 g
UN Number UN2811
Sensitivity Light sensitive
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-iodobenzoic acid