Learn More
4-Ethylphenylacetonitrile, 97%, Thermo Scientific Chemicals
Catalog Number 11343036
Click to view available options
Quantity:
1 g
5 g
51632-28-1
C10H11N
145.21
MFCD00040893
NCPKDFDQDZMXCO-UHFFFAOYSA-N
4-ethylphenylacetonitrile, 2-4-ethylphenyl acetonitrile, 4-ethylphenyl acetonitrile, benzeneacetonitrile, 4-ethyl, 2-4-ethylphenyl ethanenitrile, 4-ethylbenzylcyanide, p-ethylphenylacetonitrile, acmc-1ao1h
142867
2-(4-ethylphenyl)acetonitrile
CCC1=CC=C(CC#N)C=C1
51632-28-1
C10H11N
145.21
MFCD00040893
NCPKDFDQDZMXCO-UHFFFAOYSA-N
4-ethylphenylacetonitrile, 2-4-ethylphenyl acetonitrile, 4-ethylphenyl acetonitrile, benzeneacetonitrile, 4-ethyl, 2-4-ethylphenyl ethanenitrile, 4-ethylbenzylcyanide, p-ethylphenylacetonitrile, acmc-1ao1h
142867
2-(4-ethylphenyl)acetonitrile
CCC1=CC=C(CC#N)C=C1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 0.97
Boiling Point 110°C (1 mmHg)
Refractive Index 1.5164
Quantity 1 g
UN Number UN3276
Beilstein 2613187
Formula Weight 145.21
Percent Purity 97%
Chemical Name or Material 4-Ethylphenylacetonitrile