missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylphenoxyacetic acid, 97%, Thermo Scientific™

Product Code. 11314464
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314464 25 g 25g
11304464 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11314464 Supplier Thermo Scientific Alfa Aesar Supplier No. L01102.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24431-27-4
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00014366
InChI Key WVELHLIHMYYZAT-UHFFFAOYSA-N
Synonym 4-ethylphenoxy acetic acid, 2-4-ethylphenoxy acetic acid, 4-ethylphenoxyacetic acid, 2-p-ethylphenoxy acetic acid, acetic acid, 4-ethylphenoxy, acmc-1cdn7, p-ethylphenoxy acetic acid, cambridge id 5147400, acetic acid,2-4-ethylphenoxy
PubChem CID 90499
IUPAC Name 2-(4-ethylphenoxy)acetic acid
SMILES CCC1=CC=C(OCC(O)=O)C=C1

Specifications

Melting Point 92°C to 94°C
Quantity 25 g
Beilstein 2259506
Formula Weight 180.2
Percent Purity 97%
Chemical Name or Material 4-Ethylphenoxyacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.