Learn More
4-[(Ethoxycarbonyl)methyl]phenylboronic acid, 97%
Product Code. 11956141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
11956141 1 g 1g
1 options
Product Code. 11956141
Brand: Thermo Scientific Acros 445430010
Request Bulk or Custom Format
CAS: 92243-74-8 | C10H13BO4 | 208.02 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 92243-74-8
Melting Point 118°C to 120°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C10H13BO4
MDL Number MFCD08704683
Quantity 1 g
InChI Key RDIRUZGSOKLANG-UHFFFAOYSA-N
SMILES CCOC(=O)CC1=CC=C(C=C1)B(O)O
IUPAC Name [4-(2-ethoxy-2-oxoethyl)phenyl]boronic acid
Molecular Weight (g/mol) 208.02
Formula Weight 208.02
Percent Purity 97%
Chemical Name or Material 4-[(Ethoxycarbonyl)methyl]phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-¢(Ethoxycarbonyl)methyl!phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.