Learn More
4-Dimethylaminoazobenzene-4'-Sulfonyl Chloride, 99.5%, MP Biomedicals™
Product Code. 11486702 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486702 1 g 1g
11476702 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11486702 Supplier MP Biomedicals Supplier No. 0215093191
  • ONLINE EXCLUSIVE PRICE
    2045.00 NOK
    1944.46 NOK / 1g
    Save 100.54 NOK
    4% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Chromophoric label for amino acids, peptides, and proteins. Soluble in acetonitrile and DMF; slightly soluble in acetone
Chemical Identifiers
CAS 56512-49-3
Molecular Formula C14H14ClN3O2S
Molecular Weight (g/mol) 323.80
MDL Number MFCD00007444
InChI Key VTVWTPGLLAELLI-UHFFFAOYSA-N
Synonym dabsyl chloride, 4-dimethylaminoazobenzene-4'-sulfonyl chloride, 4-dimethylamino azobenzene-4'-sulfonyl chloride, p-dimethylaminoazobenzene-4-sulfonyl chloride, p-p-dimethylamino phenyl azo benzenesulfonyl chloride, benzenesulfonyl chloride, 4-4-dimethylamino phenyl azo, benzenesulfonyl chloride, p-p-dimethylamino phenyl azo, 4-n,n-dimethylaminoazobenzene-4'-sulfonyl chloride, p-4-dimethylamino phenyl azo benzenesulphonyl chloride
PubChem CID 91660
IUPAC Name 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonyl chloride
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(Cl)(=O)=O
Specifications
Color Red
Quantity 1 g
Formula Weight 323.8
Physical Form Crystalline Powder
Chemical Name or Material 4-Dimethylaminoazobenzene-4'-Sulfonyl Chloride