missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11486966
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486966 5 g 5g
11476966 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486966 Supplier Thermo Scientific Alfa Aesar Supplier No. B24052.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store away from incompatible materials such as oxidizing agents. Storage temperature is 2 - 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3447-53-8
Molecular Formula C8H7BrF2O
Molecular Weight (g/mol) 237.04
MDL Number MFCD00085007
InChI Key IFNMJBFIRKAQBT-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-difluoromethoxy benzene, 4-difluoromethoxy benzyl bromide, 4-difluoromethoxybenzyl bromide, 4-difluoromethoxy benzylbromide, p-difluoromethoxy benzyl bromide, benzene, 1-bromomethyl-4-difluoromethoxy, 1-bromomethyl-4-difluoromethoxy-benzene, 4-bromomethyl phenoxy difluoromethane, acmc-1aezp, p-difluoromethoxybenzylbromide
PubChem CID 2737001
IUPAC Name 1-(bromomethyl)-4-(difluoromethoxy)benzene
SMILES FC(F)OC1=CC=C(CBr)C=C1

Specifications

Boiling Point 64°C to 66°C (0.3 mmHg)
Odor Pungent
Refractive Index 1.52
Quantity 5 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.