missing translation for 'onlineSavingsMsg'
Learn More

(4-Cyanobenzyl)triphenylphosphonium chloride, 99%, Thermo Scientific Chemicals

Catalog Number p-7045840
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
20430-33-5
C26H21ClNP
413.88
MFCD00011752
GXMFPJIKPZDNCS-UHFFFAOYSA-M
4-cyanobenzyl triphenylphosphonium chloride, 4-cyanophenyl methyl triphenylphosphanium chloride, 4-triphenylphosphino methyl benzenecarbonitrile, chloride, acmc-20apna, phosphonium, 4-cyanophenyl methyl triphenyl-, chloride, 4-cyanobenzyltriphenylphosphonium chloride, p-cyanobenzyl-triphenylphosphonium chloride, 4-cyanobenzyl triphenylphosphoniumchloride, 4-cyanobenzyl-triphenylphosphonium chloride
2724468
[Cl-].N#CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
This item is not returnable. View return policy
20430-33-5
C26H21ClNP
413.88
MFCD00011752
GXMFPJIKPZDNCS-UHFFFAOYSA-M
4-cyanobenzyl triphenylphosphonium chloride, 4-cyanophenyl methyl triphenylphosphanium chloride, 4-triphenylphosphino methyl benzenecarbonitrile, chloride, acmc-20apna, phosphonium, 4-cyanophenyl methyl triphenyl-, chloride, 4-cyanobenzyltriphenylphosphonium chloride, p-cyanobenzyl-triphenylphosphonium chloride, 4-cyanobenzyl triphenylphosphoniumchloride, 4-cyanobenzyl-triphenylphosphonium chloride
2724468
[Cl-].N#CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 322°C to 326°C
Odor Odorless
Quantity 5 g
Beilstein 4630137
IUPAC Name (4-cyanophenyl)methyl-triphenylphosphanium;chloride
Formula Weight 413.89
Percent Purity 99%
Chemical Name or Material (4-Cyanobenzyl)triphenylphosphonium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'