Learn More
4-Cyanobenzoyl chloride, 98%
Description
It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is a reactant for organocatalytic tandem three component reactions of aldehyde, alkyl vinyl ketone, and amide. It is also employed as a reactant for synthesis of selective angiotensin II AT2 receptor agonists with reduced CYP 450 inhibition, allosteric glucokinase activators, hydantoin derivatives with antiproliferative activity, thiohydantoins and P2X7 receptor antagonists.
Solubility
Reacts with water.
Notes
Store under dry inert gas. Keep away from moisture, oxidizing agents, acids, bases, active metals, amines and alcohols.
Specifications
Specifications
| Melting Point | 68°C to 70°C |
| Odor | Irritating |
| Quantity | 100 g |
| UN Number | UN3261 |
| Beilstein | 386729 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Reacts with water. |
| Formula Weight | 165.58 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-Cyanobenzoyl chloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.