Learn More
4-Cyanobenzenesulfonyl chloride, 97%
Product Code. 11488016
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488016 5 g 5g
11478016 1 g 1g
11498016 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11488016 Supplier Thermo Scientific Alfa Aesar Supplier No. A14081.06
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
CAS: 49584-26-1 | C7H4ClNO2S | 201.62 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 49584-26-1
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.62
MDL Number MFCD00052337
InChI Key DBMFYTQPPBBKHI-UHFFFAOYSA-N
Synonym 4-cyanobenzene-1-sulfonyl chloride, 4-cyanobenzenesulfonylchloride, p-cyanobenzenesulphonyl chloride, 4-cyano benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyano, 4-cyanophenylsulfonyl chloride, p-cyanobenzenesulfonyl chloride, 4-cyanobenzenesulphonyl chloride, 4-cyano-benzenesulfonyl chloride, 4-chlorosulfonyl benzenecarbonitrile
PubChem CID 39497
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C#N
Spezifikation
Melting Point 107°C to 110°C
Quantity 5 g
UN Number UN3261
Beilstein 777939
IUPAC Name 4-cyanobenzenesulfonyl chloride
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzenesulfonyl chloride