Learn More
4-Chlorothiophenol(4-Chlorobenzenethiol), TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 4-Chlorothiophenol(4-Chlorobenzenethiol) |
| CAS | 106-54-7 |
| Molecular Formula | C6H5ClS |
| Quantity | 5 g |
| Synonym | 4-Chlorobenzenethiol, p-Chlorobenzenethiol, 1-Chloro-4-mercaptobenzene, 4-Chlorobenzenethiol, 4-Chlorophenylmercaptan, 4-Chlorophenylthiol, 4-Mercaptophenyl chloride, NSC 18714, NSC 229564, p-Chlorobenzenethiol, p-Chlorophenylmercaptan, p-Chlorophenylthiol, p-Chlorothiophenol, p-Mercaptochlorobenzene |
| InChI Formula | InChI=1S/C6H5ClS/c7-5-1-3-6(8)4-2-5/h1-4,8H |
| SMILES | Sc1ccc(Cl)cc1 |
| IUPAC Name | 4-chlorobenzenethiol |
| Molecular Weight (g/mol) | 144.62 |