Learn More
4-Chlorostyrene, Sigma-Aldrich™
Product Code. 15884071
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15884071 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15884071 Supplier Sigma Aldrich Supplier No. 8024580010
 Restricted Item
Estimated Shipment: 26-11-2026
MQ200 Graded - For Research and Non-regulated Industrial Applications
Stabilized with 4-tert-butylpyrocatechol for synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 1073-67-2
Density 1.09 g/cm3 at 20°C
Boiling Point 192°C
MDL Number MFCD00000632
Quantity 10 mL
Synonym 4-Chlorostyrene, 1-Chloro-4-ethenyl-bezene, 4-Chlorostyrene
Molecular Weight (g/mol) 138.59
Grade Research