Learn More
4-Chloroquinazoline, 95%, Thermo Scientific™
Product Code. 17750569 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 17750569
Brand: Thermo Scientific Acros 469400010
Request Bulk or Custom Format
Specifications
Alpha Vector CHLOROQUINAZOLINE
Chemical Name or Material 4-Chloroquinazoline
CAS 5190-68-1
Melting Point 100°C to 105°C
Color Brown to White
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C8 H5 Cl N2
MDL Number 00228682
Quantity 1 g
UN Number 2811
Molecular Weight (g/mol) 164.59
Percent Purity 0.95
Physical Form Crystals or Powder