missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorophenyl phosphorodichloridate, 98+%

Product Code. 11452907
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452907 25 g 25g
11442907 5 g 5g
11462907 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452907 Supplier Thermo Scientific Alfa Aesar Supplier No. A15507.14
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 772-79-2 | C6H4Cl3O2P | 245.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Chlorophenyl phosphorodichloridate is used as phosphorylation agent. Reactant for preparation of antimicrobial agents, antioxidant agents, antitumor agents, nucleotide anti-viral prodrug.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents, alcohols, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 772-79-2
Molecular Formula C6H4Cl3O2P
Molecular Weight (g/mol) 245.42
MDL Number MFCD00009705
InChI Key CCZMQYGSXWZFKI-UHFFFAOYSA-N
Synonym 4-chlorophenyl phosphorodichloridate, p-chlorophenyl dichlorophosphate, 4-chlorophenyl dichlorophosphate, 4-chlorophenylphosphorodichloridate, phosphorodichloridic acid, 4-chlorophenyl ester, 4-chlorophenylphosphoryl dichloride, acmc-1bjgo, cczmqygsxwzfki-uhfffaoysa, 4-chlorophenyl dichlorophosphinate, 4-chlorophenyl dichloridophosphate #
PubChem CID 69879
IUPAC Name 1-chloro-4-dichlorophosphoryloxybenzene
SMILES C1=CC(=CC=C1OP(=O)(Cl)Cl)Cl

Specifications

Density 1.508
Boiling Point 113°C to 115°C (0.9 mmHg)
Flash Point 106°C (222°F)
Odor Acrid
Refractive Index 1.5398
Quantity 25 g
UN Number UN3265
Beilstein 910754
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 245.43
Percent Purity ≥98%
Chemical Name or Material 4-Chlorophenyl phosphorodichloridate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.