missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Chloromethyl)-N-(2-fluorophenyl)benzamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
100 mg
500 mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Notes
Store away from oxidizing agents. Store in a dry, cool conditions in a well sealed containers.
Store away from oxidizing agents. Store in a dry, cool conditions in a well sealed containers.
Specifications
Quantity | 100 mg |
Formula Weight | 263.69 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Chloromethyl)-N-(2-fluorophenyl)benzamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'