Learn More
4-Chlorobenzotrifluoride, TRC
Product Code. 30367648
Change view
Click to view available options
Quantity:
1 g
500 g
Unit Size:
1g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367648 1 g 1g
30368080 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30367648 Supplier Toronto Research Chemicals Supplier No. TRCC3645251G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 98-56-6
Molecular Formula C7 H4 Cl F3
Molecular Weight (g/mol) 180.55
Synonym Toluene, p-chloro-α,α,α-trifluoro- (6CI,7CI,8CI), 1-Chloro-4-(trifluoromethyl)benzene, (p-Chlorophenyl)trifluoromethane, 4-(Trifluoromethyl)chlorobenzene, 4-(Trifluoromethyl)phenyl chloride, 4-Chloro(trifluoromethyl)benzene, 4-Chloro-1-trifluoromethylbenzene, 4-Chloro-α,α,α-benzotrifluoride, 4-Chloro-α,α,α-trifluorotoluene, 4-Chlorobenzotrifluoride, 4-Chlorobenzyltrifluoride, 4-Trifluoromethyl-1-chlorobenzene, NSC 10309, Oxol 100, Oxsol 100, Oxsul 100, p-(Trifluoromethyl)chlorobenzene, p-Chloro(trifluoromethyl)benzene, p-Chloro-α,α,α-trifluorotoluene, p-Chlorobenzotrifluoride, p-Trifluoromethylphenyl chloride
IUPAC Name 1-chloro-4-(trifluoromethyl)benzene
SMILES FC(F)(F)c1ccc(Cl)cc1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C7H4ClF3/c8-6-3-1-5(2-4-6)7(9,10)11/h1-4H
Formula Weight 179.9954
Chemical Name or Material 4-Chloro-alpha,alpha,alpha-trifluorotoluene