Learn More
4-Chlorobenzaldehyde-2,3,5,6-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chlorobenzaldehyde-2,3,5,6-d4 |
| CAS | 62285-59-0 |
| Molecular Formula | C7 2H4 H Cl O |
| Quantity | 10 mg |
| Synonym | Benzaldehyde-2,3,5,6-d4, 4-chloro- (9CI, ACI), 4-Chlorobenzaldehyde-2,3,5,6-d4 (ACI), 4-Chlorobenzaldehyde-d4, p-Chlorobenzaldehyde-d4, 4-Chlorobenzaldehyde-2,3,5,6-D4 |
| InChI Formula | InChI=1S/C7H5ClO/c8-7-3-1-6(5-9)2-4-7/h1-5H/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c(C=O)c([2H])c([2H])c1Cl |
| IUPAC Name | 4-chloro-2,3,5,6-tetradeuteriobenzaldehyde |
| Molecular Weight (g/mol) | 144.59 |
| Formula Weight | 144.028 |