missing translation for 'onlineSavingsMsg'
Learn More

4-Chloroaniline, Mikromol™

Product Code. p-200131322
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 18073910

Brand: Mikromol MM0162.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Chloroaniline
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Chloroaniline
CAS 106-47-8
Analyte or Component Names 4-Chloroaniline
Shipping Condition Room Temperature
Molecular Formula C6 H6 Cl N
Certifications/Compliance ISO 17025
Synonym Proguanil Hydrochloride Imp. B (EP), Proguanil Imp. B (EP), 4-Chloroaniline, 1-Amino-4-chlorobenzene, 4-Amino-1-chlorobenzene, 4-Aminochlorobenzene, 4-Chloro-1-aminobenzene, 4-Chlorobenzenamine, 4-Chlorophenylamine, NSC 36941, Pestanal, p-Aminochlorobenzene, p-Chloroaniline, p-Chlorophenylamine, Chlorhexidine Diacetate Imp. P (EP), Chlorhexidine Dihydrochloride Imp. P (EP), p-chloroaniline, Proguanil related, Chlorhexidine Diacetate Impurity P, Chlorhexidine Dihydrochloride Impurity P, Proguanil Hydrochloride Impurity B, Chlorhexidine Impurity P, Chlorhexidine Impurity P, Proguanil Impurity B
InChI Formula InChI=1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2
SMILES Nc1ccc(Cl)cc1
IUPAC Name 4-chloroaniline
Molecular Weight (g/mol) 127.57
Quantity 25 mg
Formula Weight 127.0189 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.