Learn More
4-Chloro-O-Tolyoxyacetic Acid, SPEX CertiPrep™
Product Code. 16350468 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
16350468 1 mL 1mL
1 options
Product Code. 16350468
Brand: SPEX Certiprep S955
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
4-Chloro-o-tolyoxyacetic acid94-74-61000μg/mL
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Packaging 2mL Amber Prescored Glass Ampule
CAS 94-74-6, 67-56-1
CAS Min % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molecular Formula C9H9ClO3
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI Key WHKUVVPPKQRRBV-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)Cl)OCC(=O)O
IUPAC Name 2-(4-chloro-2-methylphenoxy)acetic acid
Solvent or Matrix Methanol
Molecular Weight (g/mol) 200.618
PubChem CID 7204
Quantity 1 mL
ChEBI CHEBI:50099
Chemical Name or Material 4-Chloro-o-tolyoxyacetic Acid
Show More Show Less
Product intended for laboratory use only