missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-ethoxy-2-fluorobenzyl bromide, 97%, Thermo Scientific™

Catalog Number p-7052470
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1323966-30-8
C9H9BrClFO
267.522
MFCD19687186
CSQVWQCEILWYKD-UHFFFAOYSA-N
4-chloro-3-ethoxy-2-fluorobenzyl bromide, 1-bromomethyl-4-chloro-3-ethoxy-2-fluorobenzene
66523615
1-(bromomethyl)-4-chloro-3-ethoxy-2-fluorobenzene
CCOC1=C(C=CC(=C1F)CBr)Cl
This item is not returnable. View return policy
1323966-30-8
C9H9BrClFO
267.522
MFCD19687186
CSQVWQCEILWYKD-UHFFFAOYSA-N
4-chloro-3-ethoxy-2-fluorobenzyl bromide, 1-bromomethyl-4-chloro-3-ethoxy-2-fluorobenzene
66523615
1-(bromomethyl)-4-chloro-3-ethoxy-2-fluorobenzene
CCOC1=C(C=CC(=C1F)CBr)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3261
Formula Weight 267.52
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Chloro-3-ethoxy-2-fluorobenzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'