Learn More
4-Chloro-2-(trifluoromethoxy)benzylamine, 97%, Thermo Scientific™
Product Code. 15497067
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497067 250 mg 250mg
15487067 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15497067 Supplier Thermo Scientific Alfa Aesar Supplier No. H34140.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1261605-72-4
Molecular Formula C8H7ClF3NO
Molecular Weight (g/mol) 225.595
MDL Number MFCD18393826
InChI Key FCHFGYFFPMJWQT-UHFFFAOYSA-N
Synonym 4-chloro-2-trifluoromethoxy benzylamine, 1-4-chloro-2-trifluoromethoxy phenyl methanamine, 4-chloro-2-trifluoromethoxy phenyl methanamine
PubChem CID 46912354
IUPAC Name [4-chloro-2-(trifluoromethoxy)phenyl]methanamine
SMILES C1=CC(=C(C=C1Cl)OC(F)(F)F)CN
Specifications
Quantity 250 mg
UN Number UN3267
Sensitivity Air sensitive
Formula Weight 225.6
Percent Purity 97%
Chemical Name or Material 4-Chloro-2-(trifluoromethoxy)benzylamine