Learn More
Alfa Aesar™ 4'-Chloro-2'-methylacetanilide, 97%
Product Code. 11440467
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440467 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11440467 Supplier Alfa Aesar™ Supplier No. B22371.09
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 5202-86-8
Melting Point 139°C to 140°C
Molecular Formula C9H10ClNO
MDL Number MFCD00027399
Quantity 10 g
Beilstein 2087943
Synonym 4-chloro-2-methylacetanilide, n-4-chloro-2-methylphenyl acetamide, acetamide, n-4-chloro-2-methylphenyl, 4'-chloro-2'-methylacetanilide, maybridge1_000114, 4-chloro-2-methylaceta-nilide, acetamide,n-4-chloro-2-methylphenyl, n-4-chloro-2-methyl-phenyl-acetamide
InChI Key WMHHXYOPEQARIN-UHFFFAOYSA-N
SMILES CC1=C(C=CC(=C1)Cl)NC(=O)C
IUPAC Name N-(4-chloro-2-methylphenyl)acetamide
Molecular Weight (g/mol) 183.635
PubChem CID 78877
Formula Weight 183.64
Percent Purity 97%
Chemical Name or Material 4'-Chloro-2'-methylacetanilide