Learn More
4-Chloro-2-fluorophenylacetic Acid (~85%), TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-2-fluorophenylacetic Acid |
| CAS | 194240-75-0 |
| Molecular Formula | C8H6O2FCl |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C8H6ClFO2/c9-6-2-1-5(3-8(11)12)7(10)4-6/h1-2,4H,3H2,(H,11,12) |
| SMILES | O=C(O)CC1=C(F)C=C(Cl)C=C1 |
| IUPAC Name | 2-(4-chloro-2-fluorophenyl)acetic acid |
| Molecular Weight (g/mol) | 188.58 |
| Formula Weight | 188.004 |