Learn More
4-Chloro-2-fluorophenylacetic Acid (~85%), TRC
Product Code. 30367383
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30367383 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30367383 Supplier Toronto Research Chemicals Supplier No. TRCC58570850MG
 Restricted Item
Estimated Shipment: 25-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Chloro-2-fluorophenylacetic Acid
CAS 194240-75-0
Molecular Formula C8H6O2FCl
Quantity 50 mg
InChI Formula InChI=1S/C8H6ClFO2/c9-6-2-1-5(3-8(11)12)7(10)4-6/h1-2,4H,3H2,(H,11,12)
SMILES O=C(O)CC1=C(F)C=C(Cl)C=C1
IUPAC Name 2-(4-chloro-2-fluorophenyl)acetic acid
Molecular Weight (g/mol) 188.58
Formula Weight 188.004