Learn More
4-Chloro-1-napthalenecarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Chloro-1-napthalenecarboxylic Acid |
| CAS | 1013-04-3 |
| Molecular Formula | C11 H7 Cl O2 |
| Quantity | 2.5 g |
| Synonym | 4-Chloro-1-naphthalenecarboxylic Acid, 4-Chloro-1-naphthoic Acid |
| InChI Formula | InChI=1S/C11H7ClO2/c12-10-6-5-9(11(13)14)7-3-1-2-4-8(7)10/h1-6H,(H,13,14) |
| SMILES | OC(=O)c1ccc(Cl)c2ccccc12 |
| IUPAC Name | 4-chloronaphthalene-1-carboxylic acid |
| Molecular Weight (g/mol) | 206.63 |
| Formula Weight | 206.0135 |