missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1,2-difluorobenzene, 98%

Product Code. p-7020838
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499743 5 g 5g
11409753 25 g 25g
11419753 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499743 Supplier Thermo Scientific Alfa Aesar Supplier No. A12924.06
Add to basket
This item is not returnable. View return policy

CAS: 696-02-6 | C6H3ClF2 | 148.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-02-6
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00042572
InChI Key OPQMRQYYRSTBME-UHFFFAOYSA-N
Synonym 1-chloro-3,4-difluorobenzene, 3,4-difluorochlorobenzene, 4-chloro-1,2-difluoro-benzene, benzene, 4-chloro-1,2-difluoro, pubchem3417, 3,4-diflurochlorobenzene, acmc-209o9n, ksc493g2d, 3,4-difluoro-1-chlorobenzene
PubChem CID 136519
IUPAC Name 4-chloro-1,2-difluorobenzene
SMILES C1=CC(=C(C=C1Cl)F)F

Specifications

Density 1.35
Boiling Point 128°C to 130°C
Flash Point 32°C (89°F)
Odor Aromatic
Refractive Index 1.4745
Quantity 5 g
UN Number UN1993
Beilstein 2081081
Formula Weight 148.54
Percent Purity 98%
Chemical Name or Material 4-Chloro-1,2-difluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.