Learn More
4-Chloro-1,2-difluorobenzene, 98%
Product Code. 11499743
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499743 5 g 5g
11409753 25 g 25g
11419753 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11499743 Supplier Thermo Scientific Alfa Aesar Supplier No. A12924.06
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
CAS: 696-02-6 | C6H3ClF2 | 148.537 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 696-02-6
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00042572
InChI Key OPQMRQYYRSTBME-UHFFFAOYSA-N
Synonym 1-chloro-3,4-difluorobenzene, 3,4-difluorochlorobenzene, 4-chloro-1,2-difluoro-benzene, benzene, 4-chloro-1,2-difluoro, pubchem3417, 3,4-diflurochlorobenzene, acmc-209o9n, ksc493g2d, 3,4-difluoro-1-chlorobenzene
PubChem CID 136519
IUPAC Name 4-chloro-1,2-difluorobenzene
SMILES C1=CC(=C(C=C1Cl)F)F
Specifications
Density 1.35
Boiling Point 128°C to 130°C
Flash Point 32°C (89°F)
Odor Aromatic
Refractive Index 1.4745
Quantity 5 g
UN Number UN1993
Beilstein 2081081
Formula Weight 148.54
Percent Purity 98%
Chemical Name or Material 4-Chloro-1,2-difluorobenzene