Learn More
4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
Product Code. 15459735
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459735 1 g 1g
15469735 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15459735 Supplier Thermo Scientific Alfa Aesar Supplier No. H52316.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 957345-69-6
Molecular Formula C13H16BF2NO3
Molecular Weight (g/mol) 283.08
MDL Number MFCD17215212
InChI Key GSZDBQKZAWFMIC-UHFFFAOYSA-N
Synonym 4-carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzamide
PubChem CID 59023072
IUPAC Name 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=C(C(N)=O)C(F)=C1
Specifications
Quantity 1 g
Formula Weight 283.08
Percent Purity 96%
Chemical Name or Material 4-Carbamoyl-3,5-difluorobenzeneboronic acid pinacol ester