Learn More
4-Bromophenyl isothiocyanate, 97%
Product Code. 11327556
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327556 25 g 25g
11317556 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11327556 Supplier Thermo Scientific Alfa Aesar Supplier No. L11101.14
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
CAS: 1985-12-2 | C7H4BrNS | 214.08 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1985-12-2
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD00004808
InChI Key XQACWEBGSZBLRG-UHFFFAOYSA-N
Synonym 4-bromophenyl isothiocyanate, trichofytocid, p-bromophenyl isothiocyanate, 4-bromophenylisothiocyanate, benzene, 1-bromo-4-isothiocyanato, bromobenzene-4-isothiocyanate, isothiocyanic acid, p-bromophenyl ester, acmc-1bqku, wln: scnr de, p-bromophenylisothiocyanate
PubChem CID 16133
IUPAC Name 1-bromo-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)Br
Specifications
Melting Point 56°C to 60°C
Boiling Point 144°C to 145°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
UN Number UN1759
Beilstein 878549
Formula Weight 214.09
Percent Purity 97%
Chemical Name or Material 4-Bromophenyl isothiocyanate