missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobutyric acid, 97%

Product Code. 10306640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10306640 25 g Glass Bottle 25g
10123140 5 g Glass Bottle 5g
10617492 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10306640 Supplier Thermo Scientific Acros Supplier No. 133470250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2623-87-2 | C4H7BrO2 | 167.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2623-87-2
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00002817
InChI Key GRHQDJDRGZFIPO-UHFFFAOYSA-N
Synonym 4-bromobutyric acid, butanoic acid, 4-bromo, 4-bromo-n-butyric acid, butyric acid, 4-bromo, gamma-bromobutyric acid, rarechem al bo 0176, 4-bromobutyricacid, carboxypropyl bromide, 4-bromo-butyric acid, pubchem3754
PubChem CID 75809
IUPAC Name 4-bromobutanoic acid
SMILES OC(=O)CCCBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 28°C to 32°C
Color Brown
Boiling Point 301°C
Flash Point 124°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula Br(CH2)3COOH
Quantity 25 g
Beilstein 02, 283
Solubility Information Solubility in water: .. Other solubilities: soluble in dilute alkali,soluble in most common organic solvents
Formula Weight 167
Percent Purity 98%
Physical Form Low Melting Crystals
Chemical Name or Material 4-Bromobutyric acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobutyric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.