missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzenesulfonyl Chloride, TRC
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Specifications
Specifications
| Chemical Name or Material | 4-Bromobenzenesulfonyl Chloride |
| CAS | 98-58-8 |
| Molecular Formula | C6H4BrClO2S |
| Quantity | 50 g |
| Synonym | p-Bromobenzenesulfonyl Chloride, (4-Bromophenyl)chlorosulfone, 4-Bromobenzene-1-sulfonyl Chloride, 4-Bromophenylsulfonyl Chloride, 4'-Bromobenzenesulfonyl Chloride, Brosyl Chloride, NSC 4506, p-Bromophenylsulfonyl Chloride, p-Brosyl Chloride |
| InChI Formula | InChI=1S/C6H4BrClO2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H |
| SMILES | ClS(=O)(=O)c1ccc(Br)cc1 |
| IUPAC Name | 4-bromobenzenesulfonyl chloride |
| Molecular Weight (g/mol) | 255.52 |
| Formula Weight | 253.8804 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction