Learn More
4-Bromoaniline, Sigma-Aldrich™
Specifications
Specifications
| CAS | 106-40-1 |
| Melting Point | 60°C to 64°C |
| Boiling Point | 230°C to 250°C/1013 hPa |
| MDL Number | MFCD00007822 |
| Quantity | 100 g |
| Synonym | 4-Bromoaniline, p-Bromoaniline, p-Bromophenylamine, 4-Bromoaniline |
| Molecular Weight (g/mol) | 172.02 |
| Grade | Research |