Learn More
4-Bromo-N-Fmoc-L-phenylalanine, 95%
Product Code. 15428457
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15428457 5 g 5g
15438457 250 mg 250mg
15418457 1 g 1g
3 options
Product Code. 15428457
Brand: Thermo Scientific Alfa Aesar H51978.06
Request Bulk or Custom Format
CAS: 198561-04-5 | C24H20BrNO4 | 466.33 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 198561-04-5
Molecular Formula C24H20BrNO4
Molecular Weight (g/mol) 466.33
MDL Number MFCD00273460
InChI Key TVBAVBWXRDHONF-QFIPXVFZSA-N
Synonym fmoc-l-4-bromophenylalanine, s-n-fmoc-4-bromophenylalanine, fmoc-phe 4-br-oh, fmoc-4-bromo-l-phenylalanine, fmoc-l-4-br-phe-oh, fmoc-p-bromo-phe-oh, fmoc-d-4-bromophenylalanine, fmoc-l-4-bromophe, fmoc-l-phe 4-br-oh, fmoc-4'-bromo-l-phe
PubChem CID 2734449
SMILES OC(=O)[C@H](CC1=CC=C(Br)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Specifications
Melting Point 165°C to 168°C
Quantity 5 g
IUPAC Name (2S)-3-(4-bromophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Formula Weight 466.33
Percent Purity 95%
Chemical Name or Material 4-Bromo-N-Fmoc-L-phenylalanine