missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-fluorobenzyl alcohol, 96%

Product Code. p-7039997
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15485918 250 mg 250mg
15475918 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15485918

Brand: Thermo Scientific Alfa Aesar H59557.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 222978-01-0 | C7H6BrFO | 205.026 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 222978-01-0
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD08236860
InChI Key VQDUFYPJCUBGQW-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorophenyl methanol, 4-bromo-3-fluorobenzyl alcohol, 4-bromo-3-fluorobenzenemethanol, benzenemethanol, 4-bromo-3-fluoro, 4-bromo-3-fluoro-phenyl-methanol, benzenemethanol,4-bromo-3-fluoro, acmc-1cr86, ksc549c4b, 3-fluoro-4-bromo benzylalcohol, 3-fluoro-4-bromobenzyl alcohol
PubChem CID 20682297
IUPAC Name (4-bromo-3-fluorophenyl)methanol
SMILES C1=CC(=C(C=C1CO)F)Br

Specifications

Quantity 250 mg
Formula Weight 205.03
Percent Purity 96%
Chemical Name or Material 4-Bromo-3-fluorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.