Learn More
4-Bromo-3-fluorobenzyl alcohol, 96%
Product Code. 15485918
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15485918
Brand: Thermo Scientific Alfa Aesar H59557.MD
Request Bulk or Custom Format
CAS: 222978-01-0 | C7H6BrFO | 205.026 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 222978-01-0
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.026
MDL Number MFCD08236860
InChI Key VQDUFYPJCUBGQW-UHFFFAOYSA-N
Synonym 4-bromo-3-fluorophenyl methanol, 4-bromo-3-fluorobenzyl alcohol, 4-bromo-3-fluorobenzenemethanol, benzenemethanol, 4-bromo-3-fluoro, 4-bromo-3-fluoro-phenyl-methanol, benzenemethanol,4-bromo-3-fluoro, acmc-1cr86, ksc549c4b, 3-fluoro-4-bromo benzylalcohol, 3-fluoro-4-bromobenzyl alcohol
PubChem CID 20682297
IUPAC Name (4-bromo-3-fluorophenyl)methanol
SMILES C1=CC(=C(C=C1CO)F)Br
Specifications
Quantity 250 mg
Formula Weight 205.03
Percent Purity 96%
Chemical Name or Material 4-Bromo-3-fluorobenzyl alcohol