Learn More
4-Bromo-3-fluoroaniline, 98%, Thermo Scientific™
Product Code. 10224884 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10224884 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10224884 Supplier Thermo Scientific Acros Supplier No. 398250050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 656-65-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 68°C to 73°C
Color Tan to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H5BrFN
Linear Formula BrC6H3(F)NH2
MDL Number MFCD00672933
Quantity 5 g
Synonym 4-bromo-3-fluorophenylamine, 3-fluoro-4-bromoaniline, 4-bromo-3-fluoro aniline, benzenamine, 4-bromo-3-fluoro, 4-brom-3-fluoranilin, zlchem 544, pubchem1526, acmc-1badx, 4-bromo-3-fluro-aniline, 4-bromo-3-fluoro-aniline
InChI Key YTMVYYAKOPIJCZ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1N)F)Br
IUPAC Name 4-bromo-3-fluoroaniline
Molecular Weight (g/mol) 190.015
PubChem CID 821848
Formula Weight 190.01
Percent Purity ≥97.5% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-fluoroaniline
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromo-3-fluoroaniline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.