missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-(trifluoromethyl)benzaldehyde, 95%, Thermo Scientific™

Product Code. p-7029515
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15403587 250 mg 250mg
11431620 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15403587 Supplier Thermo Scientific Alfa Aesar Supplier No. H26986.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Bromo-2-(trifluoromethyl)benzaldehyde is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a pharmaceutical intermediate and intermediate in organic syntheses.

Solubility
Sparingly soluble in water (0.083 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Air Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 861928-27-0
Molecular Formula C8H4BrF3O
Molecular Weight (g/mol) 253.018
MDL Number MFCD09037472
InChI Key PROPHDFGNNBHJX-UHFFFAOYSA-N
Synonym 4-bromo-2-trifluoromethyl benzaldehyde, 5-bromo-2-formylbenzotrifluoride, 4-bromo-2-trifluoro methyl benzaldehyde, 4-bromo-alpha,alpha,alpha-trifluoro-o-tolualdehyde, pubchem18506, acmc-209q8w, 4-bromo-2-trifluoromethyl-benzaldehyde, benzaldehyde,4-bromo-2-trifluoromethyl, 5-bromo-2-formylbenzotrifluoride, 4-bromo-alpha,alpha,alpha-trifluoro-o-tolualdehyde
PubChem CID 12187218
IUPAC Name 4-bromo-2-(trifluoromethyl)benzaldehyde
SMILES C1=CC(=C(C=C1Br)C(F)(F)F)C=O

Specifications

Quantity 250 mg
Sensitivity Air Sensitive
Solubility Information Sparingly soluble in water (0.083g/L) (25°C).
Formula Weight 253.02
Percent Purity 95%
Chemical Name or Material 4-Bromo-2-(trifluoromethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.