Learn More
4-Bromo-2-fluoro-N-methylbenzamide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Bromo-2-fluoro-N-methylbenzamide |
| CAS | 749927-69-3 |
| Molecular Formula | C8H7BrFNO |
| Quantity | 2.5 g |
| Synonym | Enzalutamide Impurity 62 |
| InChI Formula | InChI=1S/C8H7BrFNO/c1-11-8(12)6-3-2-5(9)4-7(6)10/h2-4H,1H3,(H,11,12) |
| SMILES | CNC(=O)c1ccc(Br)cc1F |
| IUPAC Name | 4-bromo-2-fluoro-N-methylbenzamide |
| Molecular Weight (g/mol) | 232.05 |
| Formula Weight | 230.9695 |