missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Benzoylpiperidine hydrochloride, 98%

Product Code. 11307089
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307089 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11307089 Supplier Alfa Aesar™ Supplier No. H26887.06
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 25519-80-6
Molecular Formula C12H16ClNO
Molecular Weight (g/mol) 225.716
MDL Number MFCD00066982
InChI Key NXYKIFZJQXOUJS-UHFFFAOYSA-N
Synonym 4-benzoylpiperidine hydrochloride, phenyl piperidin-4-yl methanone hydrochloride, 4-benzoylpiperidine hcl, phenyl 4-piperidyl methanone hydrochloride, 4-benzoyl piperidine hydrochloride, 4-benzoylpiperidine hydrogen chloride, methanone, phenyl-4-piperidinyl-, hydrochloride, phenyl 4-piperidyl ketone, chloride, 4-benzoyl piperidine hcl, pubchem9881
PubChem CID 2724437
IUPAC Name phenyl(piperidin-4-yl)methanone;hydrochloride
SMILES C1CNCCC1C(=O)C2=CC=CC=C2.Cl

Specifications

Quantity 5 g
Formula Weight 225.72
Percent Purity 98%
Chemical Name or Material 4-Benzoylpiperidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.