missing translation for 'onlineSavingsMsg'
Learn More

4-Aminohippuric acid, 99%, Thermo Scientific Chemicals

Catalog Number p-3777266
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
61-78-9
194.19
MFCD00007890
HSMNQINEKMPTIC-UHFFFAOYSA-N
4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
2148
CHEBI:104011
2-[(4-aminobenzoyl)amino]acetic acid
C1=CC(=CC=C1C(=O)NCC(=O)O)N
This item is not returnable. View return policy
61-78-9
194.19
MFCD00007890
HSMNQINEKMPTIC-UHFFFAOYSA-N
4-aminohippuric acid, aminohippuric acid, p-aminohippuric acid, paha, glycine, n-4-aminobenzoyl, n-4-aminobenzoyl glycine, para-aminohippuric acid, aminohippurate, nefrotest, n-p-aminobenzoyl glycine
2148
CHEBI:104011
2-[(4-aminobenzoyl)amino]acetic acid
C1=CC(=CC=C1C(=O)NCC(=O)O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color Gray to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Molecular Formula C9H10N2O3
Linear Formula H2NC6H4CONHCH2CO2H
Quantity 10 g
Beilstein 14, II, 258
Merck Index 15, 439
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol, benzene, aceton and most, organic solvents, pract. insoluble in ccl4, very soluble in dilute hydrochloric acid and alkal
Formula Weight 194.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Aminohippuric acid
Show More Show Less
Product Identifier
  • 4-Aminohippuric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'